Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATGCATGTCAAGTTCACCTGGA[A/G]CATTGCACAGCTCTGGTAAGAACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608966 | ||||||||||||||||||||
Literature Links: |
DACT2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DACT2 - dishevelled binding antagonist of beta catenin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286350.1 | 792 | Intron | NP_001273279.1 | |||
NM_001286351.1 | 792 | Missense Mutation | GCT,GTT | A,V 224 | NP_001273280.1 | |
NM_214462.4 | 792 | Intron | NP_999627.2 | |||
XM_011535507.2 | 792 | Intron | XP_011533809.1 |