Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCTGTAAGTTCACCAGTTGTAT[C/T]ACATGATAAGGGGCAGCACTCAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606111 | ||||||||||||||||||||
Literature Links: |
MCHR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MCHR2 - melanin concentrating hormone receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040179.1 | 1122 | Silent Mutation | GTA,GTG | V,V 269 | NP_001035269.1 | |
NM_032503.2 | 1122 | Silent Mutation | GTA,GTG | V,V 269 | NP_115892.2 |