Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTAATCTCTGAGCCTGTGTTTT[A/G]ATCATCTTCATCCGACTGCTGCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603473 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LATS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LATS1 - large tumor suppressor kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270519.1 | 3755 | Intron | NP_001257448.1 | |||
NM_004690.3 | 3755 | Missense Mutation | NP_004681.1 | |||
XM_006715603.3 | 3755 | Intron | XP_006715666.1 | |||
XM_011536252.2 | 3755 | Intron | XP_011534554.1 | |||
XM_017011474.1 | 3755 | Nonsense Mutation | XP_016866963.1 | |||
XM_017011475.1 | 3755 | Nonsense Mutation | XP_016866964.1 | |||
XM_017011476.1 | 3755 | Nonsense Mutation | XP_016866965.1 | |||
XM_017011477.1 | 3755 | Intron | XP_016866966.1 | |||
XM_017011478.1 | 3755 | Nonsense Mutation | XP_016866967.1 | |||
XM_017011479.1 | 3755 | UTR 3 | XP_016866968.1 | |||
XM_017011480.1 | 3755 | Nonsense Mutation | XP_016866969.1 | |||
XM_017011481.1 | 3755 | Nonsense Mutation | XP_016866970.1 | |||
XM_017011482.1 | 3755 | Intron | XP_016866971.1 |