Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGACCAGCCAAGTGGGTCAGCACG[A/T]CCGTGGAGTCTATGGACTGGGATTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605825 | ||||||||||||||||||||
Literature Links: |
HEBP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HEBP2 - heme binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001326380.1 | 424 | Missense Mutation | ACC,TCC | T,S 63 | NP_001313309.1 | |
NM_001326381.1 | 424 | Missense Mutation | ACC,TCC | T,S 52 | NP_001313310.1 | |
NM_014320.2 | 424 | Missense Mutation | ACC,TCC | T,S 52 | NP_055135.1 | |
XM_017010631.1 | 424 | Missense Mutation | ACC,TCC | T,S 52 | XP_016866120.1 |