Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCACTGGCGCGGGCCAGGCTGGCG[A/G]TGAAGTCCAGGAGCTCGTTGCGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612808 | ||||||||||||||||||||
Literature Links: |
LRFN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRFN2 - leucine rich repeat and fibronectin type III domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020737.2 | 3246 | Missense Mutation | ACC,ATC | T,I 612 | NP_065788.1 | |
XM_011514761.2 | 3246 | Missense Mutation | ACC,ATC | T,I 612 | XP_011513063.1 | |
XM_011514762.2 | 3246 | Missense Mutation | ACC,ATC | T,I 612 | XP_011513064.1 | |
XM_017011110.1 | 3246 | Missense Mutation | ACC,ATC | T,I 612 | XP_016866599.1 | |
XM_017011111.1 | 3246 | Intron | XP_016866600.1 |