Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCTCAGAAGAAAACCATCACGC[A/G]GATTGTGAAGGATAAAAAGAAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612659 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFX6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RFX6 - regulatory factor X6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173560.3 | 398 | Missense Mutation | CAG,CGG | Q,R 112 | NP_775831.2 | |
XM_011535589.1 | 398 | Missense Mutation | CAG,CGG | Q,R 112 | XP_011533891.1 | |
XM_017010477.1 | 398 | Intron | XP_016865966.1 |