Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCTTTTACTCACGTGTTGGAGG[C/T]GCCCATTGCTACTGCCACAATGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180297 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RHAG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RHAG - Rh-associated glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000324.2 | 1119 | Missense Mutation | ACC,GCC | T,A 353 | NP_000315.2 | |
XM_011514788.1 | 1119 | Intron | XP_011513090.1 |