Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCCACACATCCAAGAAGGCAGA[C/T]GTGGTTCTGTTGTGTGGAGACCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607129 MIM: 603498 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MICAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MICAL1 - microtubule associated monooxygenase, calponin and LIM domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPIL6 - peptidylprolyl isomerase like 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMPD2 - sphingomyelin phosphodiesterase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003080.2 | 907 | Silent Mutation | GAC,GAT | D,D 171 | NP_003071.2 | |
XM_005267109.1 | 907 | UTR 5 | XP_005267166.1 | |||
XM_011536079.1 | 907 | Silent Mutation | GAC,GAT | D,D 66 | XP_011534381.1 |