Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTGCTACCTCCCTCCCCCAGTG[C/T]GTAAATATGTTGCTGTGGAGCTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610774 MIM: 606817 | ||||||||||||||||||||
Literature Links: |
CNPY3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNPY3 - canopy FGF signaling regulator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318842.1 | 595 | Missense Mutation | CGT,TGT | R,C 52 | NP_001305771.1 | |
NM_001318845.1 | 595 | Intron | NP_001305774.1 | |||
NM_001318847.1 | 595 | Missense Mutation | CGT,TGT | R,C 52 | NP_001305776.1 | |
NM_001318848.1 | 595 | Missense Mutation | CGT,TGT | R,C 52 | NP_001305777.1 | |
NM_006586.4 | 595 | Missense Mutation | CGT,TGT | R,C 52 | NP_006577.2 |
CNPY3-GNMT - CNPY3-GNMT readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318856.1 | 595 | Intron | NP_001305785.1 | |||
NM_001318857.1 | 595 | Intron | NP_001305786.1 | |||
NM_001318858.1 | 595 | Intron | NP_001305787.1 |
PTCRA - pre T-cell antigen receptor alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |