Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605660 MIM: 602306 MIM: 601962 MIM: 611440 | ||||||||||||||||||||
Literature Links: |
MIR6834 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR6834 - microRNA 6834 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6873 - microRNA 6873 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PFDN6 - prefoldin subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185181.2 | 223 | Missense Mutation | GCA,GGA | A,G 35 | NP_001172110.1 | |
NM_001265595.1 | 223 | Missense Mutation | GCA,GGA | A,G 35 | NP_001252524.1 | |
NM_001265596.1 | 223 | Missense Mutation | GCA,GGA | A,G 35 | NP_001252525.1 | |
NM_014260.3 | 223 | Missense Mutation | GCA,GGA | A,G 35 | NP_055075.1 |
RGL2 - ral guanine nucleotide dissociation stimulator like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAPBP - TAP binding protein (tapasin) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR46 - WD repeat domain 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164267.1 | 223 | Intron | NP_001157739.1 | |||
NM_005452.5 | 223 | Intron | NP_005443.3 | |||
XM_017011484.1 | 223 | Intron | XP_016866973.1 | |||
XM_017011485.1 | 223 | Intron | XP_016866974.1 | |||
XM_017011486.1 | 223 | Intron | XP_016866975.1 | |||
XM_017011487.1 | 223 | Intron | XP_016866976.1 |