Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCAGCCGGGAGGAGGCAATGGTA[C/T]GGTCCTGCCACACAGGCACAGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613976 MIM: 615660 MIM: 603170 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FANCE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FANCE - Fanconi anemia complementation group E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR7111 - microRNA 7111 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL10A - ribosomal protein L10a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEAD3 - TEA domain transcription factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003214.3 | 889 | Missense Mutation | CAT,CGT | H,R 218 | NP_003205.2 |