Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGCTGGGCTGAGCTGACACAAC[C/T]GAAGAGTGCGTCAATGGCGTTGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602265 | ||||||||||||||||||||
Literature Links: |
NEDD9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NEDD9 - neural precursor cell expressed, developmentally down-regulated 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142393.1 | 2358 | Missense Mutation | AGT,GGT | S,G 731 | NP_001135865.1 | |
NM_001271033.1 | 2358 | Missense Mutation | AGT,GGT | S,G 582 | NP_001257962.1 | |
NM_006403.3 | 2358 | Missense Mutation | AGT,GGT | S,G 731 | NP_006394.1 | |
NM_182966.3 | 2358 | Intron | NP_892011.2 |