Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTCTTCTCCCCACAGGATCGTC[G/T]CAGGAGTCGACTGGAAGCTCTGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605701 | ||||||||||||||||||||
Literature Links: |
TRIM10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIM10 - tripartite motif containing 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM15 - tripartite motif containing 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033229.2 | 868 | Missense Mutation | CGC,CTC | R,L 130 | NP_150232.2 | |
XM_011514987.1 | 868 | Missense Mutation | CGC,CTC | R,L 25 | XP_011513289.1 | |
XM_011514988.2 | 868 | UTR 5 | XP_011513290.1 |