Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTAGAACCAGGGCCGAAGCTCGCC[C/T]CTGGGTGGATGAAGACTTAAAAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612025 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IYD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IYD - iodotyrosine deiodinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164694.1 | 265 | Missense Mutation | CCC,CTC | P,L 42 | NP_001158166.1 | |
NM_001164695.1 | 265 | Missense Mutation | CCC,CTC | P,L 42 | NP_001158167.1 | |
NM_001318495.1 | 265 | UTR 5 | NP_001305424.1 | |||
NM_203395.2 | 265 | Missense Mutation | CCC,CTC | P,L 42 | NP_981932.1 | |
XM_006715478.3 | 265 | Missense Mutation | CCC,CTC | P,L 42 | XP_006715541.1 | |
XM_006715479.3 | 265 | Intron | XP_006715542.1 |