Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTGATCGTGGCAGGCGCCACCA[C/T]GCTCTTCTGCCTGCTGCACTTTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153440 MIM: 600978 MIM: 191160 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100287329 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100287329 - uncharacterized LOC100287329 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LTA - lymphotoxin alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LTB - lymphotoxin beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNF - tumor necrosis factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000594.3 | 312 | Missense Mutation | ACG,ATG | T,M 46 | NP_000585.2 |