Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTAGGAACTACCCGGACCTGTC[C/G]TTGGGAGACTACTCCCTGCTCTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120820 MIM: 613815 MIM: 600985 | ||||||||||||||||||||
Literature Links: |
C4B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C4B - complement component 4B (Chido blood group) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CYP21A2 - cytochrome P450 family 21 subfamily A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000500.7 | 434 | Silent Mutation | TCC,TCG | S,S 109 | NP_000491.4 | |
NM_001128590.3 | 434 | Silent Mutation | TCC,TCG | S,S 79 | NP_001122062.3 |
TNXB - tenascin XB | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |