Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTTCATCCTGGTCCAGTTCTGC[A/C]ACTAGTTCTTGCCTACAAACAAATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602614 | ||||||||||||||||||||
Literature Links: |
MAP3K7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP3K7 - mitogen-activated protein kinase kinase kinase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003188.3 | 1859 | Silent Mutation | GTG,GTT | V,V 524 | NP_003179.1 | |
NM_145331.2 | 1859 | Silent Mutation | GTG,GTT | V,V 551 | NP_663304.1 | |
NM_145332.2 | 1859 | Missense Mutation | GGC,TGC | G,C 513 | NP_663305.1 | |
NM_145333.2 | 1859 | Missense Mutation | GGC,TGC | G,C 486 | NP_663306.1 | |
XM_006715553.2 | 1859 | Silent Mutation | GTG,GTT | V,V 421 | XP_006715616.1 | |
XM_017011226.1 | 1859 | Silent Mutation | GTG,GTT | V,V 394 | XP_016866715.1 |