Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTATGGCTGCGACGTGGGGTCGG[A/T]CGGGCGCTTCCTCCGCGGGTACCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142800 | ||||||||||||||||||||
Literature Links: |
HLA-A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HLA-A - major histocompatibility complex, class I, A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242758.1 | 473 | Missense Mutation | GAC,GTC | D,V 130 | NP_001229687.1 | |
NM_002116.7 | 473 | Missense Mutation | GAC,GTC | D,V 130 | NP_002107.3 |