Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAATTCATTGTGTCGACTATAAG[C/T]AGAACCTTTCTGATCACCTTGATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606013 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO5 - F-box protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142522.1 | 1172 | Missense Mutation | ACT,GCT | T,A 309 | NP_001135994.1 | |
NM_012177.3 | 1172 | Missense Mutation | ACT,GCT | T,A 355 | NP_036309.1 |