Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACATAGAAGAGAAGGAACTCGATG[A/C]TTTCTTTCTCCACATGTTGATGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609202 | ||||||||||||||||||||
Literature Links: |
SCGN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCGN - secretagogin, EF-hand calcium binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006998.3 | 319 | Missense Mutation | GAT,GCT | D,A 39 | NP_008929.2 | |
XM_017010181.1 | 319 | Intron | XP_016865670.1 |