Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAATGTCTGCAGCAGACACTGA[C/T]TTAGAAACCTTTTTTAAAGCCATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606696 | ||||||||||||||||||||
Literature Links: |
GINM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GINM1 - glycoprotein integral membrane 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KATNA1 - katanin catalytic subunit A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204076.1 | 1762 | UTR 3 | NP_001191005.1 | |||
NM_007044.3 | 1762 | Silent Mutation | AAA,AAG | K,K 471 | NP_008975.1 | |
XM_005266804.2 | 1762 | Silent Mutation | AAA,AAG | K,K 471 | XP_005266861.1 | |
XM_005266805.3 | 1762 | Silent Mutation | AAA,AAG | K,K 418 | XP_005266862.1 | |
XM_017010207.1 | 1762 | Silent Mutation | AAA,AAG | K,K 471 | XP_016865696.1 | |
XM_017010208.1 | 1762 | Silent Mutation | AAA,AAG | K,K 343 | XP_016865697.1 | |
XM_017010209.1 | 1762 | Silent Mutation | AAA,AAG | K,K 343 | XP_016865698.1 | |
XM_017010210.1 | 1762 | Silent Mutation | AAA,AAG | K,K 471 | XP_016865699.1 |
RPS18P9 - ribosomal protein S18 pseudogene 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |