Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACGCCGGAGGTGAAGGTGGCTTG[C/T]TCCGAAGATGTGGACTTGCCCTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604534 | ||||||||||||||||||||
Literature Links: |
CD83 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD83 - CD83 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040280.1 | 88 | Silent Mutation | TGC,TGT | C,C 27 | NP_001035370.1 | |
NM_001251901.1 | 88 | UTR 5 | NP_001238830.1 | |||
NM_004233.3 | 88 | Silent Mutation | TGC,TGT | C,C 27 | NP_004224.1 |