Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTCCATGGTACTGGCGTTGAGC[A/C]TCCAGGGAGAAGCAGCCCAGTTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605996 MIM: 600478 MIM: 604977 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DXO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DXO - decapping exoribonuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005510.3 | 498 | Missense Mutation | GAG,GAT | E,D 55 | NP_005501.2 | |
XM_006715005.3 | 498 | Missense Mutation | GAG,GAT | E,D 55 | XP_006715068.1 | |
XM_017010329.1 | 498 | UTR 5 | XP_016865818.1 |
SKIV2L - Ski2 like RNA helicase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STK19 - serine/threonine kinase 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004197.1 | 498 | Intron | NP_004188.1 | |||
NM_032454.1 | 498 | Intron | NP_115830.1 |