Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGATAAATTTGCGTCCACATAT[G/T]GTGCACTCGTACTTCCGCCTCTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604698 MIM: 616595 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKAP12 - A-kinase anchoring protein 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB2 - zinc finger and BTB domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020861.2 | 1473 | Silent Mutation | ACA,ACC | T,T 366 | NP_065912.1 | |
XM_005267076.2 | 1473 | Silent Mutation | ACA,ACC | T,T 366 | XP_005267133.1 | |
XM_011536004.2 | 1473 | Silent Mutation | ACA,ACC | T,T 366 | XP_011534306.1 | |
XM_017011137.1 | 1473 | Silent Mutation | ACA,ACC | T,T 287 | XP_016866626.1 |