Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACTGGAATTCGGTTGCTGACCC[A/C]GCCCCAGCCTCGGCTTTCTTGTCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603632 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPS10 - ribosomal protein S10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014.4 | 526 | Intron | NP_001005.1 | |||
NM_001203245.2 | 526 | Intron | NP_001190174.1 | |||
NM_001204091.1 | 526 | Intron | NP_001191020.1 |
RPS10-NUDT3 - RPS10-NUDT3 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202470.2 | 526 | Silent Mutation | GCG,GCT | A,A 144 | NP_001189399.1 |