Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGGCATGGCACATTTATTCACC[A/G]AGACCAGAATCCCTGGGGGTAATCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 124450 MIM: 608276 | ||||||||||||||||||||
Literature Links: |
DDO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDO - D-aspartate oxidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC22A16 - solute carrier family 22 member 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033125.3 | 1296 | Silent Mutation | CTC,CTT | L,L 575 | NP_149116.2 | |
XM_005267184.3 | 1296 | Silent Mutation | CTC,CTT | L,L 405 | XP_005267241.1 | |
XM_011536204.2 | 1296 | Silent Mutation | CTC,CTT | L,L 532 | XP_011534506.1 | |
XM_011536205.2 | 1296 | UTR 3 | XP_011534507.1 | |||
XM_011536206.2 | 1296 | Silent Mutation | CTC,CTT | L,L 481 | XP_011534508.1 | |
XM_011536207.2 | 1296 | Silent Mutation | CTC,CTT | L,L 457 | XP_011534509.1 | |
XM_011536208.2 | 1296 | Silent Mutation | CTC,CTT | L,L 445 | XP_011534510.1 | |
XM_011536209.2 | 1296 | Silent Mutation | CTC,CTT | L,L 440 | XP_011534511.1 | |
XM_011536210.1 | 1296 | Intron | XP_011534512.1 | |||
XM_011536211.2 | 1296 | Silent Mutation | CTC,CTT | L,L 415 | XP_011534513.1 | |
XM_011536212.2 | 1296 | Silent Mutation | CTC,CTT | L,L 345 | XP_011534514.1 |