Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTCCTTGCCTTATTCATTTGAGA[C/T]AAACTGAGTTAGCATGATGTTGTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADGRF1 - adhesion G protein-coupled receptor F1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025048.3 | 2591 | Intron | NP_079324.2 | |||
NM_153840.3 | 2591 | Missense Mutation | ATC,GTC | I,V 907 | NP_722582.2 | |
XM_011514469.1 | 2591 | Missense Mutation | ATC,GTC | I,V 711 | XP_011512771.1 | |
XM_017010735.1 | 2591 | Missense Mutation | ATC,GTC | I,V 711 | XP_016866224.1 | |
XM_017010736.1 | 2591 | Missense Mutation | ATC,GTC | I,V 595 | XP_016866225.1 |