Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTAAACAGTCCACTAACCCTGC[C/T]GATAACTATCATCTGGCCCGGAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 123834 MIM: 609514 | ||||||||||||||||||||
Literature Links: |
CCND3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCND3 - cyclin D3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAF8 - TATA-box binding protein associated factor 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138572.2 | 96 | Silent Mutation | GCC,GCT | A,A 26 | NP_612639.2 | |
XM_005248849.2 | 96 | Silent Mutation | GCC,GCT | A,A 26 | XP_005248906.1 | |
XM_011514296.1 | 96 | Silent Mutation | GCC,GCT | A,A 26 | XP_011512598.1 | |
XM_017010241.1 | 96 | Silent Mutation | GCC,GCT | A,A 26 | XP_016865730.1 | |
XM_017010242.1 | 96 | UTR 5 | XP_016865731.1 | |||
XM_017010243.1 | 96 | UTR 5 | XP_016865732.1 | |||
XM_017010244.1 | 96 | Silent Mutation | GCC,GCT | A,A 26 | XP_016865733.1 | |
XM_017010245.1 | 96 | Silent Mutation | GCC,GCT | A,A 26 | XP_016865734.1 |