Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAAAATATTGGATGCATATCTGC[C/T]AACGCATTATCGTAACCGTGGTTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 617001 | ||||||||||||||||||||
Literature Links: |
ENPP5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ENPP5 - ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290072.1 | 1190 | Silent Mutation | TTA,TTG | L,L 345 | NP_001277001.1 | |
NM_001290073.1 | 1190 | Silent Mutation | TTA,TTG | L,L 251 | NP_001277002.1 | |
NM_021572.5 | 1190 | Silent Mutation | TTA,TTG | L,L 345 | NP_067547.1 | |
XM_005249260.3 | 1190 | Silent Mutation | TTA,TTG | L,L 345 | XP_005249317.1 | |
XM_011514786.2 | 1190 | Silent Mutation | TTA,TTG | L,L 345 | XP_011513088.1 |