Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGAGCCTACACCACCAATGTTCA[A/C]CCCTGAAGAGAGAATAGGGGAGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604995 | ||||||||||||||||||||
Literature Links: |
CRIP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRIP3 - cysteine rich protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_206922.2 | 199 | Missense Mutation | GTG,TTG | V,L 67 | NP_996805.2 | |
XM_005249103.3 | 199 | Missense Mutation | GTG,TTG | V,L 67 | XP_005249160.1 | |
XM_011514609.2 | 199 | Missense Mutation | GTG,TTG | V,L 67 | XP_011512911.1 | |
XM_017010856.1 | 199 | Intron | XP_016866345.1 | |||
XM_017010857.1 | 199 | Intron | XP_016866346.1 |
SLC22A7 - solute carrier family 22 member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |