Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGTCCTGATCGCCGGGTACATC[A/G]TCGTGTTCGTCGTGGCTCTCATTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602393 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HCRTR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HCRTR2 - hypocretin receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001526.3 | 502 | Missense Mutation | ATC,GTC | I,V 63 | NP_001517.2 | |
XM_017010798.1 | 502 | Missense Mutation | ATC,GTC | I,V 63 | XP_016866287.1 |