Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATTAACAAGGAGCATGTACTGGC[C/T]GCAGCAAAGGTAAAATCCAAATTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611264 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPW PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CENPW - centromere protein W | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012507.3 | 769 | Silent Mutation | GCC,GCT | A,A 77 | NP_001012525.1 | |
NM_001286524.1 | 769 | Silent Mutation | GCC,GCT | A,A 92 | NP_001273453.1 | |
NM_001286525.1 | 769 | Missense Mutation | CCG,CTG | P,L 64 | NP_001273454.1 | |
XM_017010845.1 | 769 | Missense Mutation | CCG,CTG | P,L 64 | XP_016866334.1 |