Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCAATGTGCAGCCCCTCATCTCC[A/G]TGGGCCTTGAGGATTCGGACGATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609577 MIM: 613475 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUL7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CUL7 - cullin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168370.1 | 5110 | Silent Mutation | CAC,CAT | H,H 1662 | NP_001161842.1 | |
NM_014780.4 | 5110 | Silent Mutation | CAC,CAT | H,H 1578 | NP_055595.2 | |
XM_005249503.2 | 5110 | Silent Mutation | CAC,CAT | H,H 1630 | XP_005249560.1 | |
XM_006715285.1 | 5110 | Silent Mutation | CAC,CAT | H,H 1614 | XP_006715348.1 | |
XM_011515019.2 | 5110 | Silent Mutation | CAC,CAT | H,H 1666 | XP_011513321.1 | |
XM_011515020.2 | 5110 | Silent Mutation | CAC,CAT | H,H 1634 | XP_011513322.1 | |
XM_011515021.1 | 5110 | Silent Mutation | CAC,CAT | H,H 869 | XP_011513323.1 | |
XM_017011533.1 | 5110 | Silent Mutation | CAC,CAT | H,H 1675 | XP_016867022.1 | |
XM_017011534.1 | 5110 | Silent Mutation | CAC,CAT | H,H 1671 | XP_016867023.1 | |
XM_017011535.1 | 5110 | Silent Mutation | CAC,CAT | H,H 1643 | XP_016867024.1 | |
XM_017011536.1 | 5110 | Silent Mutation | CAC,CAT | H,H 1623 | XP_016867025.1 | |
XM_017011537.1 | 5110 | Silent Mutation | CAC,CAT | H,H 1610 | XP_016867026.1 | |
XM_017011538.1 | 5110 | Silent Mutation | CAC,CAT | H,H 1591 | XP_016867027.1 | |
XM_017011539.1 | 5110 | Silent Mutation | CAC,CAT | H,H 1582 | XP_016867028.1 | |
XM_017011540.1 | 5110 | Intron | XP_016867029.1 |
RRP36 - ribosomal RNA processing 36 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |