Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCAAGCCGGTTCTTGCAATTAT[C/T]CAGGTAAGCCGAGAACAAGGTTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611427 | ||||||||||||||||||||
Literature Links: |
MTHFD1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTHFD1L - methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242767.1 | 191 | Silent Mutation | ATC,ATT | I,I 103 | NP_001229696.1 | |
NM_001242768.1 | 191 | Silent Mutation | ATC,ATT | I,I 37 | NP_001229697.1 | |
NM_001242769.1 | 191 | Silent Mutation | ATC,ATT | I,I 103 | NP_001229698.1 | |
NM_015440.4 | 191 | Silent Mutation | ATC,ATT | I,I 103 | NP_056255.2 | |
XM_005266907.4 | 191 | Missense Mutation | ATC,ATT | I,I 103 | XP_005266964.1 | |
XM_005266910.4 | 191 | Missense Mutation | ATC,ATT | I,I 103 | XP_005266967.1 | |
XM_005266911.4 | 191 | Missense Mutation | ATC,ATT | I,I 103 | XP_005266968.1 | |
XM_011535729.2 | 191 | Missense Mutation | ATC,ATT | I,I 103 | XP_011534031.1 | |
XM_011535730.2 | 191 | Missense Mutation | ATC,ATT | I,I 37 | XP_011534032.1 | |
XM_011535731.2 | 191 | Missense Mutation | ATC,ATT | I,I 37 | XP_011534033.1 | |
XM_011535732.2 | 191 | UTR 5 | XP_011534034.1 | |||
XM_011535733.2 | 191 | UTR 5 | XP_011534035.1 | |||
XM_011535734.2 | 191 | UTR 5 | XP_011534036.1 | |||
XM_011535737.2 | 191 | Missense Mutation | ATC,ATT | I,I 103 | XP_011534039.1 | |
XM_011535738.2 | 191 | Missense Mutation | ATC,ATT | I,I 103 | XP_011534040.1 | |
XM_017010702.1 | 191 | Missense Mutation | ATC,ATT | I,I 103 | XP_016866191.1 | |
XM_017010703.1 | 191 | Missense Mutation | ATC,ATT | I,I 103 | XP_016866192.1 | |
XM_017010704.1 | 191 | Missense Mutation | ATC,ATT | I,I 37 | XP_016866193.1 | |
XM_017010705.1 | 191 | UTR 5 | XP_016866194.1 | |||
XM_017010706.1 | 191 | UTR 5 | XP_016866195.1 | |||
XM_017010707.1 | 191 | UTR 5 | XP_016866196.1 | |||
XM_017010708.1 | 191 | Missense Mutation | ATC,ATT | I,I 103 | XP_016866197.1 |