Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAACTACAGCGATATCTGAAATCC[A/G]GGGTGGCCATCAGGAGAACAGGTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602436 | ||||||||||||||||||||
Literature Links: |
MICB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MICB - MHC class I polypeptide-related sequence B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289160.1 | 637 | Missense Mutation | AGG,GGG | R,G 166 | NP_001276089.1 | |
NM_001289161.1 | 637 | Missense Mutation | AGG,GGG | R,G 155 | NP_001276090.1 | |
NM_005931.4 | 637 | Missense Mutation | AGG,GGG | R,G 198 | NP_005922.2 |