Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGAGGCCACAGGCAGCAGCGCC[C/G]GCATCCAAACAAAAGACAGGAGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616953 MIM: 603763 MIM: 602881 MIM: 603384 | ||||||||||||||||||||
Literature Links: |
CUTA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CUTA - cutA divalent cation tolerance homolog (E. coli) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014433.2 | 625 | Missense Mutation | CCG,CGG | P,R 44 | NP_001014433.1 | |
NM_001014837.1 | 625 | Missense Mutation | CCG,CGG | P,R 2 | NP_001014837.1 | |
NM_001014838.1 | 625 | Missense Mutation | CCG,CGG | P,R 2 | NP_001014838.1 | |
NM_001014840.1 | 625 | Missense Mutation | CCG,CGG | P,R 25 | NP_001014840.1 | |
NM_015921.2 | 625 | Missense Mutation | CCG,CGG | P,R 2 | NP_057005.1 |
KIFC1 - kinesin family member C1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHF1 - PHD finger protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYNGAP1 - synaptic Ras GTPase activating protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |