Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTTTCCCCTTACGCCACTCCACG[A/G]TGATGGGGCTCTGGAGGCTGGGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615161 | ||||||||||||||||||||
Literature Links: |
HLA-DQB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HLA-DQB2 - major histocompatibility complex, class II, DQ beta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198858.1 | 718 | Missense Mutation | ACC,ATC | T,I 212 | NP_001185787.1 | |
NM_001300790.1 | 718 | Missense Mutation | ACC,ATC | T,I 212 | NP_001287719.1 | |
XM_005249051.4 | 718 | Missense Mutation | ACC,ATC | T,I 212 | XP_005249108.1 | |
XM_011514560.2 | 718 | Missense Mutation | ACC,ATC | T,I 212 | XP_011512862.1 | |
XM_011514561.2 | 718 | Missense Mutation | ACC,ATC | T,I 212 | XP_011512863.1 |
MIR3135B - microRNA 3135b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |