Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCGGCTCCTCATAGAGTCTAGA[C/T]AGAAGACCCTCCTCTCCGAGCCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605620 | ||||||||||||||||||||
Literature Links: |
IL20RA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL20RA - interleukin 20 receptor subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278722.1 | 1827 | Silent Mutation | CTA,CTG | L,L 469 | NP_001265651.1 | |
NM_001278723.1 | 1827 | Silent Mutation | CTA,CTG | L,L 407 | NP_001265652.1 | |
NM_001278724.1 | 1827 | Silent Mutation | CTA,CTG | L,L 385 | NP_001265653.1 | |
NM_014432.3 | 1827 | Silent Mutation | CTA,CTG | L,L 518 | NP_055247.3 | |
XM_006715506.2 | 1827 | Silent Mutation | CTA,CTG | L,L 385 | XP_006715569.1 | |
XM_011535904.2 | 1827 | Silent Mutation | CTA,CTG | L,L 407 | XP_011534206.1 | |
XM_017010954.1 | 1827 | Silent Mutation | CTA,CTG | L,L 519 | XP_016866443.1 | |
XM_017010955.1 | 1827 | Silent Mutation | CTA,CTG | L,L 470 | XP_016866444.1 |
NHEG1 - neuroblastoma highly expressed 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |