Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTCCCCGCTGGCCGCCGCCTCA[A/G]CTCGCTGCTTCGCGTCGGGAGGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605755 MIM: 608211 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCDC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCDC2 - doublecortin domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195610.1 | 186 | UTR 5 | NP_001182539.1 | |||
NM_016356.4 | 186 | UTR 5 | NP_057440.2 |
KAAG1 - kidney associated antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181337.3 | 186 | Silent Mutation | CAA,CAG | Q,Q 45 | NP_851854.1 |