Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACGACACTTGGGGTGGACTCAGTC[C/G]CTGCATGGAGCTCATACTTCTCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604876 | ||||||||||||||||||||
Literature Links: |
RCAN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RCAN2 - regulator of calcineurin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251973.1 | 796 | Missense Mutation | GCG,GGG | A,G 199 | NP_001238902.1 | |
NM_001251974.1 | 796 | Missense Mutation | GCG,GGG | A,G 199 | NP_001238903.1 | |
NM_005822.3 | 796 | Missense Mutation | GCG,GGG | A,G 153 | NP_005813.2 | |
XM_011514226.1 | 796 | Missense Mutation | GCG,GGG | A,G 199 | XP_011512528.1 |