Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTCTACCATTACCATTTCCCTG[C/T]ATATTCCCTCTGAACAGCAATTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605825 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HEBP2 - heme binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001326380.1 | 625 | Missense Mutation | CAT,TAT | H,Y 121 | NP_001313309.1 | |
NM_001326381.1 | 625 | Intron | NP_001313310.1 | |||
NM_014320.2 | 625 | Missense Mutation | CAT,TAT | H,Y 110 | NP_055135.1 | |
XM_017010631.1 | 625 | Intron | XP_016866120.1 |