Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGTTGCAATTTTACCTGGACAC[A/G]AAATGTTGGAACATGCATCCCATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606103 | ||||||||||||||||||||
Literature Links: |
ARMC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARMC2 - armadillo repeat containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SESN1 - sestrin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199933.1 | 1143 | Missense Mutation | CGT,TGT | R,C 350 | NP_001186862.1 | |
NM_001199934.1 | 1143 | Missense Mutation | CGT,TGT | R,C 284 | NP_001186863.1 | |
NM_014454.2 | 1143 | Missense Mutation | CGT,TGT | R,C 409 | NP_055269.1 |