Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCAGGCGGCACGGCGCGGCACC[C/T]GCCGCCTCTTCAAAGCGGCCTGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605842 | ||||||||||||||||||||
Literature Links: |
TBL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TBL2 - transducin (beta)-like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012453.2 | 994 | Silent Mutation | GCA,GCG | A,A 330 | NP_036585.1 | |
XM_006715923.3 | 994 | Silent Mutation | GCA,GCG | A,A 297 | XP_006715986.1 | |
XM_011516027.2 | 994 | Silent Mutation | GCA,GCG | A,A 165 | XP_011514329.1 |