Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCCTCCAGGAGCCCAGCTATGA[A/T]CTCCTTCTCCACAAGTAAGTGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147620 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL6 - interleukin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000600.4 | 459 | Missense Mutation | AAC,ATC | N,I 2 | NP_000591.1 | |
NM_001318095.1 | 459 | UTR 5 | NP_001305024.1 | |||
XM_005249745.4 | 459 | Missense Mutation | AAC,ATC | N,I 2 | XP_005249802.1 | |
XM_011515390.2 | 459 | Missense Mutation | AAC,ATC | N,I 2 | XP_011513692.1 |
LOC541472 - uncharacterized LOC541472 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |