Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCGTGCTCCGCAGGCCCTCCAG[C/G]TCTAGGGACCTGAGCATGGCCAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614506 MIM: 602576 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRAT1 - BRCA1 associated ATM activator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152743.3 | 2571 | Missense Mutation | GAC,GAG | D,E 780 | NP_689956.2 | |
XM_005249643.4 | 2571 | Missense Mutation | GAC,GAG | D,E 840 | XP_005249700.1 | |
XM_011515177.2 | 2571 | Missense Mutation | GAC,GAG | D,E 868 | XP_011513479.1 | |
XM_011515178.1 | 2571 | Missense Mutation | GAC,GAG | D,E 868 | XP_011513480.1 | |
XM_011515179.2 | 2571 | Missense Mutation | GAC,GAG | D,E 867 | XP_011513481.1 | |
XM_011515181.2 | 2571 | Missense Mutation | GAC,GAG | D,E 808 | XP_011513483.1 | |
XM_011515182.2 | 2571 | Missense Mutation | GAC,GAG | D,E 868 | XP_011513484.1 | |
XM_011515183.1 | 2571 | Missense Mutation | GAC,GAG | D,E 693 | XP_011513485.1 | |
XM_011515184.2 | 2571 | Missense Mutation | GAC,GAG | D,E 693 | XP_011513486.1 | |
XM_011515186.2 | 2571 | UTR 3 | XP_011513488.1 | |||
XM_017011833.1 | 2571 | Missense Mutation | GAC,GAG | D,E 839 | XP_016867322.1 | |
XM_017011834.1 | 2571 | Missense Mutation | GAC,GAG | D,E 779 | XP_016867323.1 | |
XM_017011835.1 | 2571 | Missense Mutation | GAC,GAG | D,E 605 | XP_016867324.1 | |
XM_017011836.1 | 2571 | UTR 3 | XP_016867325.1 |
LFNG - LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |