Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTGAACATTCTCATTCATGGGT[C/T]GATTTTCTTGGGCCACTAGGTTTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HERPUD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HERPUD2 - HERPUD family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022373.4 | 1219 | Missense Mutation | CAA,CGA | Q,R 251 | NP_071768.3 | |
XM_006715765.1 | 1219 | Missense Mutation | CAA,CGA | Q,R 187 | XP_006715828.1 | |
XM_011515473.1 | 1219 | Missense Mutation | CAA,CGA | Q,R 187 | XP_011513775.1 | |
XM_017012506.1 | 1219 | Missense Mutation | CAA,CGA | Q,R 251 | XP_016867995.1 |