Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGACACAGGGCCCCCAGTATTGA[C/T]GCACTCATAAGAACCTGCAAGTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611700 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SVOPL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SVOPL - SVOP like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139456.1 | 1059 | Silent Mutation | GCA,GCG | A,A 456 | NP_001132928.1 | |
NM_174959.3 | 1059 | Silent Mutation | GCA,GCG | A,A 304 | NP_777619.1 | |
XM_005250143.3 | 1059 | Silent Mutation | GCA,GCG | A,A 456 | XP_005250200.1 | |
XM_011515797.2 | 1059 | Silent Mutation | GCA,GCG | A,A 304 | XP_011514099.1 | |
XM_017011746.1 | 1059 | Silent Mutation | GCA,GCG | A,A 365 | XP_016867235.1 | |
XM_017011747.1 | 1059 | Silent Mutation | GCA,GCG | A,A 304 | XP_016867236.1 | |
XM_017011748.1 | 1059 | Silent Mutation | GCA,GCG | A,A 304 | XP_016867237.1 |