Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGTTTCAGCTAGACTGATCACG[A/G]TGACATTGCTCAGTCCCATGAGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611986 MIM: 610337 | ||||||||||||||||||||
Literature Links: |
MRPS24 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPS24 - mitochondrial ribosomal protein S24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
URGCP - upregulator of cell proliferation | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077663.2 | 2843 | Missense Mutation | ACC,ATC | T,I 785 | NP_001071131.1 | |
NM_001077664.2 | 2843 | Missense Mutation | ACC,ATC | T,I 742 | NP_001071132.1 | |
NM_001290075.1 | 2843 | Missense Mutation | ACC,ATC | T,I 742 | NP_001277004.1 | |
NM_001290076.1 | 2843 | Missense Mutation | ACC,ATC | T,I 742 | NP_001277005.1 | |
NM_017920.4 | 2843 | Missense Mutation | ACC,ATC | T,I 776 | NP_060390.3 |
URGCP-MRPS24 - URGCP-MRPS24 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204871.1 | 2843 | Intron | NP_001191800.1 |