Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGACCCTCCTGGATGCTCACCACC[A/G]TCACTGTGCCCGCAGGGTTCACCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602372 | ||||||||||||||||||||
Literature Links: |
ZAN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZAN - zonadhesin (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003386.2 | 655 | Missense Mutation | ATC,GTC | I,V 164 | NP_003377.2 | |
NM_173059.2 | 655 | Missense Mutation | ATC,GTC | I,V 164 | NP_775082.2 | |
XM_011516555.2 | 655 | Missense Mutation | ATC,GTC | I,V 164 | XP_011514857.1 | |
XM_017012581.1 | 655 | Intron | XP_016868070.1 |