Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTCTCCCTGGATGACTCGGTGG[A/G]TGAGACGGAGGCCGTCAAGCGCTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100740 MIM: 614469 MIM: 602933 MIM: 611481 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACHE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACHE - acetylcholinesterase (Cartwright blood group) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRRT - serrate, RNA effector molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128852.1 | 759 | Missense Mutation | GAT,GGT | D,G 164 | NP_001122324.1 | |
NM_001128853.1 | 759 | Missense Mutation | GAT,GGT | D,G 164 | NP_001122325.1 | |
NM_001128854.1 | 759 | Missense Mutation | GAT,GGT | D,G 164 | NP_001122326.1 | |
NM_015908.5 | 759 | Missense Mutation | GAT,GGT | D,G 164 | NP_056992.4 | |
XM_005250405.2 | 759 | Missense Mutation | GAT,GGT | D,G 171 | XP_005250462.1 | |
XM_005250406.2 | 759 | Missense Mutation | GAT,GGT | D,G 171 | XP_005250463.1 | |
XM_005250407.2 | 759 | Missense Mutation | GAT,GGT | D,G 171 | XP_005250464.1 | |
XM_005250408.2 | 759 | Missense Mutation | GAT,GGT | D,G 171 | XP_005250465.1 | |
XM_017012290.1 | 759 | Missense Mutation | GAT,GGT | D,G 85 | XP_016867779.1 | |
XM_017012291.1 | 759 | Missense Mutation | GAT,GGT | D,G 85 | XP_016867780.1 |
TRIP6 - thyroid hormone receptor interactor 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UFSP1 - UFM1 specific peptidase 1 (inactive) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |